Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri-Tert-Butyl Borate

Catalog No. 23660850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-660-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-660-850G Supplier Sigma Aldrich Supplier No. 23660850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tri-Tert-Butyl Borate

Specifications

CAS 7397-43-5
Density 1.005 g/mL at 25°C
Boiling Point 107°C to 110°C (23 mmHg, lit.)
Molecular Formula C4H10O2
Refractive Index n20/D 1.389
Linear Formula CH3CH(OH)CH2CH2OH
Quantity 50 g
MDL Number MFCD00064277
Synonym Boron tert-butoxide; Tri-tert-butoxyborane
Molecular Weight (g/mol) 90.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.