Alfa Aesar™ Tri-tert-butyl borate, 97%

Manufacturer: Alfa Aesar™ H5546106

Quantity 5g

 View more versions of this product

Catalog No. AAH5546106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7397-43-5
    Linear Formula [(CH3)3CO]3B
    Formula Weight 230.16
    MDL Number MFCD00009652
    UN Number UN1993
    Synonym Boric acid tri-tert-butyl ester; Boron tert-butoxide
    Chemical Name or Material Tri-tert-butyl borate

    Specifications

    Formula Weight 230.16
    Melting Point 18° to 19°C
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Physical Form Liquid
    CAS 7397-43-5
    Refractive Index 1.3890
    Color Colorless
    MDL Number MFCD00009652
    Synonym Boric acid tri-tert-butyl ester; Boron tert-butoxide
    Density 0.811g/mL
    Boiling Point 101°C (74mmHg)
    Quantity 5g
    DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Linear Formula [(CH3)3CO]3B
    Chemical Name or Material Tri-tert-butyl borate
    Assay 98%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit