Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-tert-butylphosphine, 98%

Catalog No. AA1017806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1017806 5 g
AA1017803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1017806 Supplier Thermo Scientific Chemicals Supplier No. 01017806
  • $713.50
    $418.65 / Each of 1

    Save $294.85

    41% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13716-12-6 | C12H27P | 202.32 g/mol

Tri-tert-butylphosphine is used primarily in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-12-6
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00015006
InChI Key BWHDROKFUHTORW-UHFFFAOYSA-N
Synonym tri-tert-butylphosphine, tri-t-butylphosphine, tri-tert-butylphosphane, tri-tertbutylphosphine, tris 1,1-dimethylethyl phosphine, tri-tert-butylphosphin, p t-bu 3, tri-tert-butyl-phosphane, tris tert-butyl phosphine, phosphine, tris 1,1-dimethylethyl
PubChem CID 83681
IUPAC Name tri-tert-butylphosphane
SMILES CC(C)(C)P(C(C)(C)C)C(C)(C)C

Specifications

Melting Point 30°C
Color White
Boiling Point 102°C to 103°C (13 mmHg)
Flash Point -16°C (-3°F)
Odor Pungent
Linear Formula [(CH3)3C]3P
Quantity 5 g
UN Number UN2846
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water. Soluble in alcohol and organic solvents
Formula Weight 202.32
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Tri-tert-butylphosphine
Show More Show Less
Hazard Category H228-H250-H314-H335
Hazard Statement GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 237-266-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.