Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri-Tert-Butylphosphine Solution

Catalog No. 65532550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-532-550ML 50 mL
65-532-525ML 25 mL
65-532-510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 65-532-550ML Supplier Sigma Aldrich Supplier No. 65532550ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tri-tert-butylphosphine is a suitable ligand for palladium-catalyzed coupling reactions such as:Heck ReactionsNegishi cross-couplingStille reactionsSonogashira reactions

Specifications

CAS 13716-12-6
Density 0.861 g/mL at 25°C
Molecular Formula C12H27P
Linear Formula C12H27P
MDL Number MFCD00015006
Quantity 50 mL
Synonym P(t-Bu)3; P(t-Bu)3
Molecular Weight (g/mol) 202.32 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.