Learn More
Medchemexpress LLC Tri-tert-butylphosphine tetrafluoroborate | 131274-22-1 | 98.0% | 290.13 | 25 G
SDP
Catalog No. 5000402870
Encompass_Preferred
Tri-tert-butylphosphine tetrafluoroborate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Used as a ligand in the palladium-catalyzed enantioselective alfa-arylation of N-boc-pyrrolidine.
  • Used with a palladium(0)-15-membered, triolefinic, macrocycle in Suzuki cross-coupling reactions of aryl bromides and chlorides.
  • Utilized in the heck coupling of vinyl tosylates with olefins.
Catalog No. 50-004-02870 Supplier Medchemexpress LLC Supplier No. 25GHY34451
Tri-tert-butylphosphine tetrafluoroborate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Used as a ligand in the palladium-catalyzed enantioselective alfa-arylation of N-boc-pyrrolidine.
  • Used with a palladium(0)-15-membered, triolefinic, macrocycle in Suzuki cross-coupling reactions of aryl bromides and chlorides.
  • Utilized in the heck coupling of vinyl tosylates with olefins.