Learn More
Selleck Chemical LLC Triacetin S4581-500mg
SDP
Catalog No. 503108677
Encompass_Preferred
Triacetin (Glycerol triacetate Glyceryl triacetate Glycerin triacetate 1 2 3-Triacetoxypropane) is a triglyceride that is used as an antifungal agent
Catalog No. 50-310-8677 Supplier Selleck Chemical LLC Supplier No. S4581500MG
Triacetin (Glycerol triacetate Glyceryl triacetate Glycerin triacetate 1 2 3-Triacetoxypropane) is a triglyceride that is used as an antifungal agent