Learn More
Sigma Aldrich Fine Chemicals Biosciences Triacetin 99 fCC fG25KG
Catalog No. 502902823
Encompass
Triacetin is a triester of glycerin and acetic acid that occurs naturally in papaya. It is mainly used as a synthetic flavoring agent in ice-creams nonalcoholic beverages and baked goods.
Catalog No. 50-290-2823 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W20070025KGK
Triacetin is a triester of glycerin and acetic acid that occurs naturally in papaya. It is mainly used as a synthetic flavoring agent in ice-creams nonalcoholic beverages and baked goods.