Learn More
Medchemexpress LLC Triacetin (Glyceryl triacetate) | 102-76-1 | MFCD00008716 | 99.9% | 5 G
SDP
Catalog No. 5000409262
Encompass_Preferred
Triacetin (Glyceryl triacetate) is a synthetic, orally active compound that serves as a triester of glycerol and acetic acid. It is known to increase acetate bioavailability in glioma cells, leading to growth arrest and apoptosis.
  • Freely crosses the blood-brain barrier/plasma membrane
  • Increases histone acetylation
  • Enhances the chemotherapeutic efficacy of Temozolomide (TMZ)
Catalog No. 50-004-09262 Supplier Medchemexpress LLC Supplier No. HYB08965G
Triacetin (Glyceryl triacetate) is a synthetic, orally active compound that serves as a triester of glycerol and acetic acid. It is known to increase acetate bioavailability in glioma cells, leading to growth arrest and apoptosis.
  • Freely crosses the blood-brain barrier/plasma membrane
  • Increases histone acetylation
  • Enhances the chemotherapeutic efficacy of Temozolomide (TMZ)