Learn More
Medchemexpress LLC Triacetonamine | 826-36-8 | MFCD00005975 | 155.24 | 50 G
SDP
Catalog No. 5000414787
Encompass_Preferred
Triacetonamine (2,2,6,6-Tetramethyl-4-piperidone) is a chemical compound primarily used as an intermediate in various synthesis processes. It plays a role in the production of pharmaceutical products, pesticides, and photostabilizers for polymers.
  • Intermediate for pharmaceutical product synthesis.
  • Intermediate for pesticide synthesis.
  • Intermediate for photostabilizer synthesis for polymers.
Catalog No. 50-004-14787 Supplier Medchemexpress LLC Supplier No. HYN113150G
Add to Cart
Add to Cart
Triacetonamine (2,2,6,6-Tetramethyl-4-piperidone) is a chemical compound primarily used as an intermediate in various synthesis processes. It plays a role in the production of pharmaceutical products, pesticides, and photostabilizers for polymers.
  • Intermediate for pharmaceutical product synthesis.
  • Intermediate for pesticide synthesis.
  • Intermediate for photostabilizer synthesis for polymers.