Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triacetonamine-d17 (2,2,6,6-tetramethyl-4-piperidone-d17) | 52168-48-6 | 99.7% | 172.34 | C9D17NO | 10 MG
SDP

Catalog No. 5000296575
Encompass_Preferred

Triacetonamine-d17 is the deuterium-labeled analog of triacetonamine provided for laboratory research use as an isotopic tracer or internal standard in analytical methods. It is intended for research use only and should be handled according to applicable safety procedures due to reported biological activity in animal models.

  • deuterium-labeled tracer for analytical quantitation (NMR, GC-MS, LC-MS).
  • 99.7% purity by specification.
  • molecular weight 172.34 g/mol.
  • cas number 52168-48-6 for unambiguous identification.
  • chemical formula C9D17NO.
  • packaged as 10 mg quantity.
  • store powder at -20°C; in solvent store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-96575 Supplier Medchemexpress LLC Supplier No. HYN1131S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triacetonamine-d17 is the deuterium-labeled analog of triacetonamine provided for laboratory research use as an isotopic tracer or internal standard in analytical methods. It is intended for research use only and should be handled according to applicable safety procedures due to reported biological activity in animal models.

  • deuterium-labeled tracer for analytical quantitation (NMR, GC-MS, LC-MS).
  • 99.7% purity by specification.
  • molecular weight 172.34 g/mol.
  • cas number 52168-48-6 for unambiguous identification.
  • chemical formula C9D17NO.
  • packaged as 10 mg quantity.
  • store powder at -20°C; in solvent store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.