Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetyl-beta-cyclodextrin 97.0+%, TCI America™
SDP

Catalog No. T184425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T184425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T184425G Supplier TCI America Supplier No. T184425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23739-88-0
Molecular Formula C84H112O56
Molecular Weight (g/mol) 2017.764
MDL Number MFCD00074981
InChI Key NOPKOJDDVCBPTP-DJSZNTTKSA-N
Synonym beta-Cyclodextrin Henicosaacetate
PubChem CID 71311472
SMILES CC(=O)OCC1C2C(C(C(O1)OC3C(OC(C(C3OC(=O)C)OC(=O)C)OC4C(OC(C(C4OC(=O)C)OC(=O)C)OC5C(OC(C(C5OC(=O)C)OC(=O)C)OC6C(OC(C(C6OC(=O)C)OC(=O)C)OC7C(OC(C(C7OC(=O)C)OC(=O)C)OC8C(OC(O2)C(C8OC(=O)C)OC(=O)C)COC(=O)C)COC(=O)C)COC(=O)C)COC(=O)C)COC(=O)C)COC(=O)C)OC(=O)C)O

Specifications

Color White
Quantity 25 g
Formula Weight 2017.76
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triacetyl-beta-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.