Learn More
AdipoGen Triacetyl-resveratrol
SDP
Supplier:  AdipoGen CDXA0350M050
Encompass_Preferred
Chemical. CAS 42206-94-0. Formula C20H18O6. MW 354.35. Synthetic. A cell-permeable triacetate resveratrol prodrug that is easily converted to resveratrol by esterase activity and exhibits similar bioactivity as resveratrol in cell cultures. The compound induces p53 activity and inhibits proliferation in breast and prostate tumor cell lines.
Catalog No. 50-256-1743