Learn More
AdipoGen Triacetyl-resveratrol
SDP
Catalog No. 502561744
Encompass_Preferred
Chemical. CAS 42206-94-0. Formula C20H18O6. MW 354.35. Synthetic. A cell-permeable triacetate resveratrol prodrug that is easily converted to resveratrol by esterase activity and exhibits similar bioactivity as resveratrol in cell cultures. The compound induces p53 activity and inhibits proliferation in breast and prostate tumor cell lines.
Catalog No. 50-256-1744 Supplier AdipoGen Supplier No. CDXA0350M010
Add to Cart
Add to Cart
Chemical. CAS 42206-94-0. Formula C20H18O6. MW 354.35. Synthetic. A cell-permeable triacetate resveratrol prodrug that is easily converted to resveratrol by esterase activity and exhibits similar bioactivity as resveratrol in cell cultures. The compound induces p53 activity and inhibits proliferation in breast and prostate tumor cell lines.