Learn More
Medchemexpress LLC Triacetylresveratrol | 42206-94-0 | 354.35 | 100 MG
SDP
Catalog No. 5000415296
Encompass_Preferred
Triacetylresveratrol is an acetylated analog of Resveratrol. It has anticancer effects by decreasing the phosphorylation of STAT3 and NF-κB in a dose- and time-dependent manner in PANC-1 and BxPC-3 cells.
  • Decreases the phosphorylation of STAT3 and NF-κB in PANC-1 and BxPC-3 cells.
  • Exhibits anticancer effects.
  • Significantly down-regulates anti-apoptotic Bcl-2 family protein Mcl-1.
  • Up-regulates pro-apoptotic Bcl-2 family proteins Bim and Puma.
  • Inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner.
Catalog No. 50-004-15296 Supplier Medchemexpress LLC Supplier No. HYN1410100MG
Triacetylresveratrol is an acetylated analog of Resveratrol. It has anticancer effects by decreasing the phosphorylation of STAT3 and NF-κB in a dose- and time-dependent manner in PANC-1 and BxPC-3 cells.
  • Decreases the phosphorylation of STAT3 and NF-κB in PANC-1 and BxPC-3 cells.
  • Exhibits anticancer effects.
  • Significantly down-regulates anti-apoptotic Bcl-2 family protein Mcl-1.
  • Up-regulates pro-apoptotic Bcl-2 family proteins Bim and Puma.
  • Inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner.