Learn More
Medchemexpress LLC Triacetylresveratrol | 42206-94-0 | 354.35 | 250 MG
SDP
Catalog No. 5000389476
Encompass_Preferred
Triacetylresveratrol is an acetylated analog of Resveratrol. It decreases the phosphorylation of STAT3 and NF-κB in a dose- and time-dependent manner in PANC-1 and BxPC-3 cells, demonstrating anticancer effects. It significantly down-regulates the anti-apoptotic Bcl-2 family protein Mcl-1 and up-regulates the pro-apoptotic Bcl-2 family proteins Bim and Puma. Triacetylresveratrol inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner.
  • STAT3/NF-κB inhibitor
  • Decreases the phosphorylation of STAT3 and NF-κB
  • Exhibits anticancer effects
  • Significantly down-regulates the anti-apoptotic Bcl-2 family protein Mcl-1
  • Up-regulates the pro-apoptotic Bcl-2 family proteins Bim and Puma
  • Inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner
Catalog No. 50-003-89476 Supplier Medchemexpress LLC Supplier No. 250MGHYN1410
Triacetylresveratrol is an acetylated analog of Resveratrol. It decreases the phosphorylation of STAT3 and NF-κB in a dose- and time-dependent manner in PANC-1 and BxPC-3 cells, demonstrating anticancer effects. It significantly down-regulates the anti-apoptotic Bcl-2 family protein Mcl-1 and up-regulates the pro-apoptotic Bcl-2 family proteins Bim and Puma. Triacetylresveratrol inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner.
  • STAT3/NF-κB inhibitor
  • Decreases the phosphorylation of STAT3 and NF-κB
  • Exhibits anticancer effects
  • Significantly down-regulates the anti-apoptotic Bcl-2 family protein Mcl-1
  • Up-regulates the pro-apoptotic Bcl-2 family proteins Bim and Puma
  • Inhibits cell viability and induces apoptosis of pancreatic cancer cells in a concentration and incubation time-dependent manner