Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacontane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015023 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5023 250 mg
11-101-5022 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5023 Supplier MilliporeSigma Supelco Supplier No. 90270250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triacontane is an acyclic linear alkane that physically appears like a paraffin like solid, which occurs like a waxy coat on tobacco leaves.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 638-68-6
Physical Form Neat
Linear Formula CH3(CH2)28 CH3
MDL Number MFCD00009410
Melting Point 64°C to 67°C; 64°C to 67°C
Boiling Point 258°C to 259°C (3 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 422.81
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.