Learn More
Cayman Chemical Triacsin C, 76896-80-5, 1 mg
SDP
Supplier:  Cayman Chemical 100074481
Encompass_Preferred
Molecular Formula C11H17N3O, Purity ≥95%, Formula Weight 207.3, Synonyms: FR 900190, WS 1228A; An inhibitor of long fatty acid acyl-CoA synthetase (IC50 = 6.3 M in Raji cells) that interferes with lipid metabolism by inhibiting the de novo synthesis of triglycerides, diglycerides, and cholesterol esters; acts as a hypotensive vasodilator.
Catalog No. NC0784596