Learn More
Apexbio Technology LLC Triacsin C,500ug CAS# 76896-80-5
SDP
Catalog No. 502240104
Encompass_Preferred
Triacsin C is a potent inhibitor of long chain acyl-CoA synthetase, with an IC50 value of 6.3 μM in Raji cell membrane fraction. Long chain acyl-CoA synthetase is essential for catalyzing the conversion of free fatty acids to acyl-CoAs. Inhibition of acyl-CoA synthetase has been shown to interfere lipid synthesis as well as cell proliferation in Raji lymphoma cells. Triacsin C also induces apoptosis in many cancer cell lines, such as human lung cancer cells. In addition, Triacsin C has been found to be highly effective against rotavirus replication.  Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0104 Supplier Apexbio Technology LLC Supplier No. B70700.5
Triacsin C is a potent inhibitor of long chain acyl-CoA synthetase, with an IC50 value of 6.3 μM in Raji cell membrane fraction. Long chain acyl-CoA synthetase is essential for catalyzing the conversion of free fatty acids to acyl-CoAs. Inhibition of acyl-CoA synthetase has been shown to interfere lipid synthesis as well as cell proliferation in Raji lymphoma cells. Triacsin C also induces apoptosis in many cancer cell lines, such as human lung cancer cells. In addition, Triacsin C has been found to be highly effective against rotavirus replication.  Other sizes are also available. Please inqury us for quote.