Learn More
Enzo Life Sciences Triacsin C, (100µg), CAS Number: 76896-80-5
SDP
Catalog No. 501031775
Encompass_Preferred
Acyl-CoA synthetase inhibitor. Analog of polyunsaturated fatty acid. Potent inhibitor of long-chain fatty acyl CoA synthetase. Selectively inhibits arachidonoyl-CoA synthetase in intact cells and the nonspecific acyl-CoA synthetase in cell sonicates IC50=3.6-8.7µM. Inhibits neutrophil functions. Blocks β-cell apoptosis induced by fatty acids (lipoapoptosis) and synthesis of triglycerides, diglycerides and cholesterol esters. Potent vasodilator. Alternative Name: 2,4,7-Undecatrienal nitrosohydrazone, 2E,4E,7E-Undecatriene-1-triazene, WS1228A. Formula: C11H17N3O. MW: 207.3. CAS Number: 76896-80-5. Purity: ≥90% (TLC). Appearance: Liquid. In DMSO. Solubility: Soluble in DMSO (25mg/ml) or methanol (5mg/ml). Use/Stability: Stable for up to 1 year after receipt when stored at -20°C. Long Term Storage: -20°C
Catalog No. 50-103-1775 Supplier Enzo Life Sciences Supplier No. BMLEI2180100
Acyl-CoA synthetase inhibitor. Analog of polyunsaturated fatty acid. Potent inhibitor of long-chain fatty acyl CoA synthetase. Selectively inhibits arachidonoyl-CoA synthetase in intact cells and the nonspecific acyl-CoA synthetase in cell sonicates IC50=3.6-8.7µM. Inhibits neutrophil functions. Blocks β-cell apoptosis induced by fatty acids (lipoapoptosis) and synthesis of triglycerides, diglycerides and cholesterol esters. Potent vasodilator. Alternative Name: 2,4,7-Undecatrienal nitrosohydrazone, 2E,4E,7E-Undecatriene-1-triazene, WS1228A. Formula: C11H17N3O. MW: 207.3. CAS Number: 76896-80-5. Purity: ≥90% (TLC). Appearance: Liquid. In DMSO. Solubility: Soluble in DMSO (25mg/ml) or methanol (5mg/ml). Use/Stability: Stable for up to 1 year after receipt when stored at -20°C. Long Term Storage: -20°C