Learn More
Cayman Chemical Triadimefon, Purity: ≥98%, CAS: 43121-43-3, 10 mg
SDP
Catalog No. 502980139
Encompass_Preferred
A triazole fungicide that inhibits lanosterol 14α-demethylase, interfering with ergosterol biosynthesis, and blocks gibberellin biosynthesis
Catalog No. 50-298-0139 Supplier Cayman Chemical Supplier No. 1871410 MG
A triazole fungicide that inhibits lanosterol 14α-demethylase, interfering with ergosterol biosynthesis, and blocks gibberellin biosynthesis