Learn More
Medchemexpress LLC Triadimefon | 43121-43-3 | MFCD00055506 | 98.1% | 293.75 | C14H16ClN3O2 | 250MG
SDP
Catalog No. 5000238786
Encompass_Preferred
Triadimefon is an orally active fungicide supplied as a research-grade small molecule for laboratory studies. It modulates kinase signaling and energy-sensing pathways and is used in fungal growth assays, toxicology, and developmental biology research.
  • Orally active fungicide used in research.
  • Reduces phosphorylation of AKT1 and ERK1/2.
  • Increases pAMPK levels.
  • Inhibits growth of Saccharomyces cerevisiae.
  • Shown to disrupt hormone homeostasis and affect fetal adrenal development in animal studies.
  • Induces metabolic shifts in hepatocytes and impairs spatial learning in models.
  • Available as solid and ready-to-use solution formats for different workflows.
  • High purity suitable for analytical and biological assays.
Catalog No. 50-002-38786 Supplier Medchemexpress LLC Supplier No. HY123037250MG
Triadimefon is an orally active fungicide supplied as a research-grade small molecule for laboratory studies. It modulates kinase signaling and energy-sensing pathways and is used in fungal growth assays, toxicology, and developmental biology research.
  • Orally active fungicide used in research.
  • Reduces phosphorylation of AKT1 and ERK1/2.
  • Increases pAMPK levels.
  • Inhibits growth of Saccharomyces cerevisiae.
  • Shown to disrupt hormone homeostasis and affect fetal adrenal development in animal studies.
  • Induces metabolic shifts in hepatocytes and impairs spatial learning in models.
  • Available as solid and ready-to-use solution formats for different workflows.
  • High purity suitable for analytical and biological assays.