Learn More
Cayman Chemical Triadimenol, Purity: ≥95% (mixture of isomers), CAS: 55219-65-3, 250 mg
SDP
Catalog No. 502936557
Encompass_Preferred
A metabolite of triadimefon; induces cranial nerve and ganglia abnormalities in a rat post-implantation whole embryo culture model from 12.5-125 μM; induces embryotoxicity in rats and rabbits when administered orally at doses of 100 and 40 mg/kg respectively; induces embryonic mortality as well as decreases fertility and increases the number of female offspring in medaka fish (O. latipes) from 3-3,000 μg/L
Catalog No. 50-293-6557 Supplier Cayman Chemical Supplier No. 24216250 MG
A metabolite of triadimefon; induces cranial nerve and ganglia abnormalities in a rat post-implantation whole embryo culture model from 12.5-125 μM; induces embryotoxicity in rats and rabbits when administered orally at doses of 100 and 40 mg/kg respectively; induces embryonic mortality as well as decreases fertility and increases the number of female offspring in medaka fish (O. latipes) from 3-3,000 μg/L