Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Triadine Solution
SDP

Catalog No. 10000712 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
8 oz.
16 oz.
1 gal.
Includes:
Case of 12
Case of 24
Case of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
10-000-712 1 gal. Case of 4
10-000-709 8 oz. Case of 24
10-000-710 16 oz. Case of 12
10-000-707 16 oz. Case of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-712 Supplier Stoelting™ Supplier No. 50999
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stoelting™ Triadine Solution is a non-irritating, non-staining, fast acting, antimicrobial solution.

This antiseptic germicide kills all gram positive and negative bacteria, fungi, viruses, protozoa and spores. Ideal for surgical site preparation, venipuncture, catheter exit site care and care of scrapes, minor burns, cuts and skin wounds.

Specifications

For Use With (Application) Neuroscience
Size 1 gal.
Includes Case of 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.