Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro BNP Calibration Verification Set – 2 Sets/5T each
SDP

Catalog No. 1398206
Change view
Click to view available options
Quantity:
2 Sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-982-06 2 Sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-06 Supplier Quidel Supplier No. 98015XR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triage BNP Calibration Verification materials are to be used with the Triage BNP Test and Triage Meter to verify the calibration of the Triage BNP Test throughout the measurable range.

  • EDTA human plasma containing preservatives and B-type natriuretic peptide (BNP).
  • The concentrations and standard deviations are printed on the enclosed Expected Values card.

Specifications

Content And Storage <-20°C
Concentration Varies, Across linear range
Detectable Analytes BNP
For Use With (Equipment) Triage, BNP, MeterPro
Sample Type Plasma
Includes Control, Value Assignment Sheet
Level A to E
Quantity 2 Sets
Stability 1 hour, once thawed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.