Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro BNP Controls Level 2 - 5T
SDP

Catalog No. 1398205
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-982-05 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-05 Supplier Quidel Supplier No. 98014XR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BNP Controls are assayed materials to be used with the Triage BNP Test and the Triage Meter to assist the end user in monitoring product performance.

  • EDTA human plasma containing preservatives and B-type natriuretic peptide (BNP).
  • The concentrations and standard deviations are printed on the enclosed Expected Values card.

Specifications

Content And Storage <-20°C
Concentration High Control
Detectable Analytes BNP
For Use With (Equipment) Triage, BNP, MeterPro
Sample Type Plasma
Includes Control, Value Assignment Sheet, Code Chip
Level 2
Quantity 5 Vials
Stability 1 hour, once thawed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.