Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro BNP Panel 25/PK
SDP

Catalog No. 1398203
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-982-03 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-03 Supplier Quidel Supplier No. 98000XR
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triage BNP is a rapid, point-of-care CLIA-Waived fluorescence immunoassay used with the Triage MeterPro. It is used to measure B-type natriuretic peptide (BNP) in EDTA anticoagulated whole blood or plasma specimens.

  • Sample type: Plasma or whole blood in EDTA
  • CLIA-Waived
  • Results within 15 minutes

Specifications

Content And Storage 2°C to 8°C, 14 days of room temperature
CPT Code 83880
Clia Complexity Waived, Moderate
Detectable Analytes BNP
For Use With (Application) Triage, BNP, MeterPro
Includes Test Devices, Code Chip, Paper Roll, Package Insert, Pipettes
Packaging Type Kit of BNP
Quantity 25
Range 5 - 5000 pg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.