Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro Cardiac Panel Troponin 25/PK
SDP

Catalog No. 1398210
Change view
Click to view available options
Clia Complexity:
Moderate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
13-982-10 Moderate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-10 Supplier Quidel Supplier No. 97021HS
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of troponin I in EDTA anticoagulated whole blood or plasma specimens. This can be used as an aid in the diagnosis of myocardial infarction (injury).

  • Sample type: Plasma or whole blood in EDTA
  • Results within 20 minutes

Specifications

Content And Storage 2°C to 8°C, 14 days at Room Temperature
CPT Code 84484
Clia Complexity Moderate
Detectable Analytes Troponin I
For Use With (Application) Triage, Cardiac, Troponin I, MeterPro
Includes Test Devices, Code Chip, Paper Roll, Package Insert, Pipettes
Packaging Type Kit of Cardiac - TnI only
Quantity 25 Cardiac - TnI only Test Devices
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.