Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro Combo Troponin, Troponin/CK-MB with BNP, and D-Dimer 75 total tests
SDP

Catalog No. 1398215
Change view
Click to view available options
Clia Complexity:
Moderate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
13-982-15 Moderate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-15 Supplier Quidel Supplier No. 97022BD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

See Cardiac Panel (97022HS), BNP Test and D-Dimer Test for more detail

  • Sample type: Plasma or whole blood in EDTA
  • Results within 20 minutes

Specifications

Content And Storage 2°C to 8°C, 14 days at Room Temperature
CPT Code 83880, 82553, 85379,84484
Clia Complexity Moderate
Detectable Analytes BNP, CK-MB, D-Dimer, Troponin I
For Use With (Application) Triage, BNP, Cardiac - CKMB and TnI, D-Dimer, MeterPro
Includes Test Devices, Code Chip, Paper Roll, Package Insert, Pipettes
Packaging Type Kit of BNP, Kit of Cardiac - CKMB and TnI, Kit of D-Dimer
Quantity 25 BNP Test Devices, 25 Cardiac - CLMB and TnI, 25 D-Dimer Test Devices
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.