Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro D-Dimer Panel 25/PK
SDP

Catalog No. 1398216
Change view
Click to view available options
Quantity:
25 D-Dimer Test Devices
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-982-16 25 D-Dimer Test Devices
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-16 Supplier Quidel Supplier No. 98100
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triage D-Dimer Test is a rapid, quantitative immunoassay to aid in assessing and evaluating patients with suspected disseminated intravascular coagulation (DIC) and thromboembolic events including pulmonary embolism (PE) and deep vein thrombosis (DVT).

  • Sample type: Plasma or whole blood in EDTA
  • Results within 20 minutes

Specifications

Content And Storage 2°C to 8°C, 14 days at Room Temperature
CPT Code 85379
Clia Complexity Moderate
Detectable Analytes D-dimer
For Use With (Application) Triage, D-Dimer, MeterPro
Includes Test Devices, Code Chip, Paper Roll, Package Insert, Pipettes
Packaging Type Kit of D-Dimer
Quantity 25 D-Dimer Test Devices
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.