Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage MeterPro Total Controls Level 1 - includes BNP, D DIMER, CK-MB, MYO, TNI - 5 Vials per box
SDP

Catalog No. 1398217
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-982-17 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-17 Supplier Quidel Supplier No. 88753
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triage Total 5 Controls are assayed materials to be used with the Triage Cardiac Panel, Triage BNP Test, Triage D-Dimer Test, and Triage Meters to assist the laboratory in monitoring test performance.

EDTA human plasma containing preservatives, CK-MB, myoglobin, troponin I, B-type natriuretic peptide (BNP) and D-dimer.

Specifications

Content And Storage <-20°C
Detectable Analytes Brain Natriuretic Peptide (BNP), Creatinine Kinase-MB (CK-MB), D-dimer, Myoglobin, Troponin I
For Use With (Equipment) Triage, BNP, D-Dimer, Cardiac, MeterPro
Sample Type Plasma
Includes Control, Value Assignment Sheet, Code Chip
Level A to E
Quantity 5 Vials
Stability 1 hour once thawed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.