Learn More
Ready America™ Triage Tarps
SDP
Catalog No. 19777010
Change view
Click to view available options
Color:
Red, Yellow, Green, Black
Catalog No. Color
19-777-010 Red, Yellow, Green, Black
1 options
This item is not returnable. View return policy
Catalog No. 19-777-010 Supplier Ready America™ Supplier No. 74104
Add to Cart
Add to Cart
This item is not returnable. View return policy
Life saving supplies for 3 days. This 2-person 3-day emergency kit was designed to offer additional items to those that want their emergency kit to go the extra mile.
These Ready America™ triage tarps measure 16 ft. x 20 ft. and are made of 5 mil thick vinyl with grommets and come in four colors. Sold in sets of four only. Also available with carry bag (TR07-KT).
Specifications
Product Type Emergency Supplies
Color Red, Yellow, Green, Black
Material Polyester, Vinyl
Disposable Reusable