Learn More
Medchemexpress LLC Triallate | 2303-17-5 | 99.0% | 250 MG
SDP
Catalog No. 5000233376
Encompass_Preferred
Triallate is a thiocarbamate-class pre-emergence herbicide provided as a high-purity analytical standard for research and pesticide analysis. It is supplied in a 250 mg packaged quantity and is intended for use as a reference material in residue analysis and method development.
  • High purity suitable for analytical applications.
  • Intended as a reference standard for pesticide residue testing.
  • Solid under typical laboratory conditions (appearance: white to off-white).
  • Molecular formula C10H16Cl3NOS; molecular weight 304.66 g/mol.
  • Store and handle according to SDS and COA instructions.
Catalog No. 50-002-33376 Supplier Medchemexpress LLC Supplier No. HY119435250MG
Triallate is a thiocarbamate-class pre-emergence herbicide provided as a high-purity analytical standard for research and pesticide analysis. It is supplied in a 250 mg packaged quantity and is intended for use as a reference material in residue analysis and method development.
  • High purity suitable for analytical applications.
  • Intended as a reference standard for pesticide residue testing.
  • Solid under typical laboratory conditions (appearance: white to off-white).
  • Molecular formula C10H16Cl3NOS; molecular weight 304.66 g/mol.
  • Store and handle according to SDS and COA instructions.