Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl 1,3,5-Benzenetricarboxylate 98.0+%, TCI America™
SDP

Catalog No. T12035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12035G Supplier TCI America Supplier No. T12035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 17832-16-5
Color White-Yellow
Boiling Point 212°C
Molecular Formula C18H18O6
Quantity 5 g
MDL Number MFCD00008647
Synonym triallyl trimesate, triallyl benzene-1,3,5-tricarboxylate, triallyl 1,3,5-benzenetricarboxylate, triallylbenzene-1,3,5-tricarboxylate, 1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester, 1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester, 1,3,5-benzenetricarboxylic acid triallyl ester, 1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate, triallyltrimesate, acmc-1c8hf
InChI Key VOSUIKFOFHZNED-UHFFFAOYSA-N
SMILES C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
IUPAC Name tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
Molecular Weight (g/mol) 330.336
PubChem CID 87328
Formula Weight 330.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Triallyl 1,3,5-Benzenetricarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.