Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 99%, stabilized with 100 ppm BHT, 98%, Thermo Scientific™

Catalog No. AC151295000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC151295000 500 g
AC151291000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151295000 Supplier Thermo Scientific Chemicals Supplier No. 151295000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025-15-6
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
InChI Key KOMNUTZXSVSERR-UHFFFAOYSA-N
Synonym triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
PubChem CID 13931
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Color Colorless to Yellow
Density 1.159g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Quantity 500 g
Beilstein 26, IV, 637
Formula Weight 249.27
Percent Purity ≥98.5%
Physical Form Liquid After Melting
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.