Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triallylamine

Catalog No. T45004250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T45004250ML 250 mL
T450041L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T45004250ML Supplier Sigma Aldrich Supplier No. T45004250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triallylamine (taa) reacts with primary aromatic amines in the presence of a ruthenium catalyst to form 2-ethyl-3-methylquinolines
  • Taa undergoes hydrozirconation followed by transmetalation with germanium tetrachloride to form 1-aza-5-germa-5-chlorobicyclo[3.3.3]undecane. this compound can react with grignard or lithium reagents to form the corresponding 5-organo compounds
  • The cycloaddition of taa to fluorinated 1,3,4-oxadiazoles affords octahydro-2,7-methanofuro[3,2-c]pyridines

Specifications

CAS 102-70-5
Density 0.79 g/mL (at 25°C (literature))
Boiling Point 150°C to 151°C (lit.)
Molecular Formula C9H15N
Refractive Index n20/D 1.451 (literature)
Linear Formula (H2C=CHCH2)3 N
MDL Number MFCD00026093
Quantity 250 mL
Synonym TAA
Molecular Weight (g/mol) 137.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.