Learn More
Cayman Chemical Triamcinolone acetonide, Purity: ≥98%, CAS: 76-25-5, 100 mg
SDP
Catalog No. 502966972
Encompass_Preferred
A synthetic corticosteroid; decreases cytokine levels, the firing rate of sensory neurons, and mechanical hypersensitivity in a rat spinal nerve ligation model at 1.5 mg/kg administered prior to and following surgery for three days; decreases outflow facility in a mouse model of steroid-induced glaucoma when 20 µl of a 40 mg/ml suspension is administered subconjunctivally
Catalog No. 50-296-6972 Supplier Cayman Chemical Supplier No. 18026100 MG
A synthetic corticosteroid; decreases cytokine levels, the firing rate of sensory neurons, and mechanical hypersensitivity in a rat spinal nerve ligation model at 1.5 mg/kg administered prior to and following surgery for three days; decreases outflow facility in a mouse model of steroid-induced glaucoma when 20 µl of a 40 mg/ml suspension is administered subconjunctivally