Learn More
Medchemexpress LLC Triamcinolone | 124-94-7 | 99.82% | 500 MG
SDP
Catalog No. 5000406706
Encompass_Preferred
Triamcinolone is a long-acting corticosteroid that exerts anti-inflammatory, anti-oedematous, anti-proliferative, anti-angiogenetic, immunomodulatory, and neuroprotective effects by binding to glucocorticoid receptors. It is used to relieve various dermatitis, immune diseases, and ocular diseases.
  • Long-acting corticosteroid
  • Anti-inflammatory effects
  • Anti-oedematous effects
  • Anti-proliferative effects
  • Anti-angiogenetic effects
  • Immunomodulatory effects
  • Neuroprotective effects
  • Binds to glucocorticoid receptors
  • Relieves various dermatitis
  • Relieves immune diseases
  • Relieves ocular diseases
Catalog No. 50-004-06706 Supplier Medchemexpress LLC Supplier No. HYB0328500MG
Triamcinolone is a long-acting corticosteroid that exerts anti-inflammatory, anti-oedematous, anti-proliferative, anti-angiogenetic, immunomodulatory, and neuroprotective effects by binding to glucocorticoid receptors. It is used to relieve various dermatitis, immune diseases, and ocular diseases.
  • Long-acting corticosteroid
  • Anti-inflammatory effects
  • Anti-oedematous effects
  • Anti-proliferative effects
  • Anti-angiogenetic effects
  • Immunomodulatory effects
  • Neuroprotective effects
  • Binds to glucocorticoid receptors
  • Relieves various dermatitis
  • Relieves immune diseases
  • Relieves ocular diseases