Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Triamcinolone

Catalog No. AC462240010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462240010 1 g
AC462240050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462240010 Supplier Thermo Scientific Chemicals Supplier No. 462240010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-94-7
Molecular Formula C21H27FO6
Molecular Weight (g/mol) 394.43
InChI Key GFNANZIMVAIWHM-OBYCQNJPSA-N
PubChem CID 31307
ChEBI CHEBI:9667

Specifications

Melting Point 262°C to 263°C
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf
Formula Weight 394.43
Chemical Name or Material Triamcinolone
Hazard Category GHS Signal Word: Warning
EINECSNumber 204-718-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.