Learn More
Apexbio Technology LLC Triamcinolone 124-94-7 50mg
SDP
Catalog No. 502240763
Encompass_Preferred
Triamcinolone (124-94-7) is a small-molecule agonist targeting intracellular glucocorticoid receptors (GR) It is designed to modulate the transcriptional regulation of genes involved in inflammation immune responses cellular proliferation vascular permeability and angiogenesis thereby regulating multiple cellular and immunological pathways Triamcinolone exerts its biological activity primarily through binding to GR and modulating downstream gene expression In cell-based GR assays Triamcinolone demonstrates potent receptor binding with reported IC50 values typically ranging from 0 5 to 5 nM Based on these pharmacological properties Triamcinolone holds research potential in experimental models of dermatological inflammation ophthalmic inflammation autoimmune conditions and investigations of GR-mediated signal transduction and immunoregulatory pathways
This item is not returnable. View return policy
Catalog No. 50-224-0763 Supplier Apexbio Technology LLC Supplier No. B185950
Triamcinolone (124-94-7) is a small-molecule agonist targeting intracellular glucocorticoid receptors (GR) It is designed to modulate the transcriptional regulation of genes involved in inflammation immune responses cellular proliferation vascular permeability and angiogenesis thereby regulating multiple cellular and immunological pathways Triamcinolone exerts its biological activity primarily through binding to GR and modulating downstream gene expression In cell-based GR assays Triamcinolone demonstrates potent receptor binding with reported IC50 values typically ranging from 0 5 to 5 nM Based on these pharmacological properties Triamcinolone holds research potential in experimental models of dermatological inflammation ophthalmic inflammation autoimmune conditions and investigations of GR-mediated signal transduction and immunoregulatory pathways