Learn More
Apexbio Technology LLC Triamcinolone Acetonide,1g CAS# 76-25-5
SDP
Catalog No. 502240323
Encompass_Preferred
Triamcinolone acetonide is an antiinflammatory steroid that inhibits the IgE-dependent release of histamine. This is done by human basophils and the growth of NEL-M1 human melanoma cells. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0323 Supplier Apexbio Technology LLC Supplier No. A85401000
Triamcinolone acetonide is an antiinflammatory steroid that inhibits the IgE-dependent release of histamine. This is done by human basophils and the growth of NEL-M1 human melanoma cells. Other sizes are also available. Please inqury us for quote.