Learn More
Apexbio Technology LLC Triamcinolone Acetonide 76-25-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061850
Encompass_Preferred
Triamcinolone acetonide is a synthetic glucocorticoid steroid characterized by potent anti-inflammatory activity Its molecular structure allows it to penetrate cells effectively facilitating modulation of inflammatory processes by suppressing IgE-dependent histamine release from human basophils This effect interferes with mast cell-mediated allergic response pathways reducing inflammation in affected tissues Additionally Triamcinolone acetonide inhibits proliferation of specific human cell lines including the NEL-M1 melanoma cells making it particularly useful in oncology research settings involving cell growth regulation It is widely applied in biomedical investigation for studying inflammatory signaling pathways immune cell functions and cellular proliferation mechanisms relevant to dermatological autoimmune and tumor-associated inflammatory conditions
Catalog No. 50-000-61850 Supplier Apexbio Technology LLC Supplier No. A85405.1
Triamcinolone acetonide is a synthetic glucocorticoid steroid characterized by potent anti-inflammatory activity Its molecular structure allows it to penetrate cells effectively facilitating modulation of inflammatory processes by suppressing IgE-dependent histamine release from human basophils This effect interferes with mast cell-mediated allergic response pathways reducing inflammation in affected tissues Additionally Triamcinolone acetonide inhibits proliferation of specific human cell lines including the NEL-M1 melanoma cells making it particularly useful in oncology research settings involving cell growth regulation It is widely applied in biomedical investigation for studying inflammatory signaling pathways immune cell functions and cellular proliferation mechanisms relevant to dermatological autoimmune and tumor-associated inflammatory conditions