Learn More
Medchemexpress LLC Triamcinolone acetonide | 76-25-5 | 99.95% | 250 MG
SDP
Catalog No. 5000405606
Encompass_Preferred
Triamcinolone acetonide inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells. It reduces chondrocyte viability and leads to cartilage destruction. It also activates macrophages with anti-inflammatory characteristics and can be used in the study of diseases such as atopic dermatitis.
  • Decreases proliferation of BRECs
  • Reduces chondrocyte viability in normal and osteoarthritic chondrocytes
  • Increases severity of cartilage structural damage in osteoarthritic cartilage
  • Strongly induces monocyte differentiation towards an M2 and anti-inflammatory macrophage phenotype
  • Enhances folate receptor beta (FRβ)-related macrophage activation
  • Prevents osteophyte development in an in vivo rat model of osteoarthritis
Catalog No. 50-004-05606 Supplier Medchemexpress LLC Supplier No. 250MGHYB0636R
Triamcinolone acetonide inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells. It reduces chondrocyte viability and leads to cartilage destruction. It also activates macrophages with anti-inflammatory characteristics and can be used in the study of diseases such as atopic dermatitis.
  • Decreases proliferation of BRECs
  • Reduces chondrocyte viability in normal and osteoarthritic chondrocytes
  • Increases severity of cartilage structural damage in osteoarthritic cartilage
  • Strongly induces monocyte differentiation towards an M2 and anti-inflammatory macrophage phenotype
  • Enhances folate receptor beta (FRβ)-related macrophage activation
  • Prevents osteophyte development in an in vivo rat model of osteoarthritis