Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone acetonide, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111015454 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5454 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5454 Supplier MilliporeSigma Supelco Supplier No. T65015G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 76-25-5
Physical Form Neat
Linear Formula C24H31 FO6
MDL Number MFCD00056834
Synonym 9 alpha-Fluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide; 9 alpha-Fluoro-16 alpha-hydroxyprednisolone 16 alpha,17 alpha-acetonide
Type Analytical Standard
Quantity 5 g
Formula Weight 434.50
Percent Purity ≥99%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.