Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone acetonide, Analytical Standard, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111015455 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5455 250 mg
11-101-5453 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5455 Supplier MilliporeSigma Supelco Supplier No. T6501250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. T6501.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 76-25-5
Physical Form Neat
Linear Formula C24H31FO6
MDL Number MFCD00056834
Synonym 9 alpha-Fluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide; 9 alpha-Fluoro-16 alpha-hydroxyprednisolone 16 alpha,17 alpha-acetonide
Quantity 250 mg
Formula Weight 434.50
Percent Purity ≥99%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.