Learn More
Medchemexpress LLC Triamcinolone acetonide-d6 | 352431-33-5 | 95.7% | 5 MG
SDP
Catalog No. 5000409039
Encompass_Preferred
Triamcinolone acetonide-d6 is deuterium labeled Triamcinolone acetonide. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09039 Supplier Medchemexpress LLC Supplier No. HYB0636S35MG
Triamcinolone acetonide-d6 is deuterium labeled Triamcinolone acetonide. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.