Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone Acetonide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015456 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5456 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5456 Supplier MilliporeSigma Supelco Supplier No. PHR17011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Triamcinolone Acetonide is a glucocorticoid and a derivative of pregnane, used mainly in the treatment of rheumatic disease, allergic and dermatologic symptoms.
TRUSTED_SUSTAINABILITY

Specifications

CAS 76-25-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H31 FO6
MDL Number MFCD00056834
Synonym Triamcinolone acetonide; 9 alpha-Fluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide; 9 alpha-Fluoro-16 alpha-hydroxyprednisolone 16 alpha,17 alpha-acetonide
Type Certified Reference Material
Quantity 1 g
Formula Weight 434.50
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.