Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone Acetonide, Powder, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18607512 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-512 5 g Amber Glass Bottle
18-607-513 10 g Poly Bottle
18-607-516 25 g Amber Glass Bottle
18-607-514 100 g Poly Bottle
18-607-515 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-512 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR1075GM
Add to Cart
Add to Cart

TR107, 76-25-5, C24H31FO6

Spectrum™ Chemical Triamcinolone Acetonide, Powder, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 76-25-5
Molecular Formula C24H31FO6
Molecular Weight (g/mol) 434.50
InChI Key YNDXUCZADRHECN-JNQJZLCISA-N
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

CAS Max % 1
Loss on Drying 0.015
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation +118° to +130°
Percent Purity 97 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.