Learn More
Medchemexpress LLC Triamcinolone hexacetonide | 5611-51-8 | 532.64 | 25 MG
SDP
Catalog No. 5000448073
Encompass_Preferred
Triamcinolone hexacetonide is a commonly used, long-acting steroid for the treatment of subacute and chronic inflammatory joint diseases. This compound is intended for research use only.
  • Used in the treatment of subacute and chronic inflammatory joint diseases.
  • Shows a dose-dependent protective effect against chemically induced articular cartilage damage.
  • Reduces chondrocyte cloning, osteophyte formation, and fibrillation in animal models.
  • Local application may reduce mechanical sensitivity and enhance regeneration after nerve injury.
  • High purity of 98.06%.
  • Appears as a white to off-white solid.
  • Soluble in DMSO for in vitro applications.
Catalog No. 50-004-48073 Supplier Medchemexpress LLC Supplier No. 25MGHYU00103
Triamcinolone hexacetonide is a commonly used, long-acting steroid for the treatment of subacute and chronic inflammatory joint diseases. This compound is intended for research use only.
  • Used in the treatment of subacute and chronic inflammatory joint diseases.
  • Shows a dose-dependent protective effect against chemically induced articular cartilage damage.
  • Reduces chondrocyte cloning, osteophyte formation, and fibrillation in animal models.
  • Local application may reduce mechanical sensitivity and enhance regeneration after nerve injury.
  • High purity of 98.06%.
  • Appears as a white to off-white solid.
  • Soluble in DMSO for in vitro applications.