Learn More
Medchemexpress LLC Triamcinolone (standard) | 124-94-7 | 98.02% | 25 MG
SDP
Catalog No. 5000450987
Encompass_Preferred
Triamcinolone (Standard) is the analytical standard of Triamcinolone, intended for research and analytical applications. It is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Used as a reference standard for assays
  • Suitable for HPLC, GC, and MS techniques
  • Molecular weight of 394.43 and chemical formula C₂₁H₂₇FO₆
  • Appears as a solid with a white to off-white color
  • Classified as a steroid
  • Initial source from plants in the Solanaceae family
Catalog No. 50-004-50987 Supplier Medchemexpress LLC Supplier No. 25MGHYB0328R
Triamcinolone (Standard) is the analytical standard of Triamcinolone, intended for research and analytical applications. It is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Used as a reference standard for assays
  • Suitable for HPLC, GC, and MS techniques
  • Molecular weight of 394.43 and chemical formula C₂₁H₂₇FO₆
  • Appears as a solid with a white to off-white color
  • Classified as a steroid
  • Initial source from plants in the Solanaceae family