Learn More
Medchemexpress LLC HY-140249 100mg Medchemexpress, Tri(Amino-PEG3-amide)-amine CAS: Purity:>98%
SDP
Catalog No. 502081550
Encompass_Preferred
Medchemexpress, HY-140249 100mg Tri(Amino-PEG3-amide)-amine CAS: Tri(Amino-PEG3-amide)-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1550 Supplier Medchemexpress LLC Supplier No. HY140249100MG
Medchemexpress, HY-140249 100mg Tri(Amino-PEG3-amide)-amine CAS: Tri(Amino-PEG3-amide)-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.